Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Clec4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005880.1 | NP_001005880.1 | 4 | 423 | ||
GenBank mRNA | AY397670.1 | - | 4 | 423 | ||
AY494061.1 | - | 4 | 424 | |||
AY581052.1 | - | 4 | 423 |