Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vezt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006984.1 | NP_001006985.1 | 1, 2 | 91 | ||
XM_006241252.2 | XP_006241314.1 | 1, 2 | 150 | |||
GenBank mRNA | BC083840.1 | - | 1, 2 | 91 | 7114280 | |
FQ211922.1 | - | 1, 2 | 72 |