Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007007.1 | NP_001007008.1 | 7 | 535 | ||
XM_008770762.1 | XP_008768984.1 | 7 | 2608 | |||
XM_008770763.1 | XP_008768985.1 | 7 | 2608 | |||
XM_008770764.1 | XP_008768986.1 | 7 | 2610 | |||
XM_008770765.1 | XP_008768987.1 | 7 | 2608 | |||
GenBank mRNA | BC083791.1 | - | 7 | 535 | 7106440 |