Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007176.4 | NP_001007177.1 | 5 | 583 | ||
NM_001256596.1 | NP_001243525.1 | 5 | 558 | |||
NM_001256597.1 | NP_001243526.1 | 5 | 666 | |||
NM_001256598.1 | NP_001243527.1 | 5 | 1308 | |||
GenBank mRNA | BC017981.1 | - | 5 | 576 | 4245342 |