Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007576.2 | NP_001007577.1 | 20 | 6234 | ||
GenBank mRNA | AY651761.1 | - | 20 | 6229 |