Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007705.1 | NP_001007706.1 | 11 | 2822 | ||
XM_006240475.2 | XP_006240537.1 | 12 | 2938 | |||
XM_006240476.2 | XP_006240538.1 | 12 | 2935 | |||
GenBank mRNA | BC081966.1 | - | 11 | 2822 | 7125402 |