Gene Symbol: Mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007709.1 | NP_001007710.1 | 6 | 1133 | ||
XM_006242081.2 | XP_006242143.1 | 6 | 1186 | |||
XM_006242083.2 | XP_006242145.1 | 7 | 2192 | |||
XM_008765717.1 | XP_008763939.1 | 6 | 1319 | |||
XM_008765718.1 | XP_008763940.1 | 7 | 2946 | |||
GenBank mRNA | BC083593.1 | - | 6 | 1133 | 7132737 |