Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007741.1 | NP_001007742.1 | 2 | 947 | ||
GenBank mRNA | BC079314.1 | - | 2 | 947 | 7134903 |