Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007756.1 | NP_001007757.1 | 3 | 225 | ||
GenBank mRNA | BC079160.1 | - | 3 | 225 | 7129380 |