Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008317.1 | NP_001008318.1 | 9 | 896 | ||
GenBank mRNA | AY158662.1 | - | 9 | 896 | ||
FQ211910.1 | - | 990 | ||||
FQ216126.1 | - | 9 | 951 | |||
FQ227188.1 | - | 9 | 957 | |||
FQ234126.1 | - | 9 | 956 | |||
FQ234429.1 | - | 9 | 996 |