Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008331.1 | NP_001008332.1 | 10 | 1073 | ||
XM_006245190.2 | XP_006245252.1 | 11 | 1168 | |||
GenBank mRNA | BC085807.1 | - | 10 | 1073 | 7112886 |