Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008380.1 | NP_001008381.1 | 13 | 2352 | ||
XM_006243261.2 | XP_006243323.1 | 13 | 2455 | |||
XM_006243262.2 | XP_006243324.1 | 13 | 2259 | |||
XM_006243263.2 | XP_006243325.1 | 13 | 2784 | |||
XM_008766311.1 | XP_008764533.1 | 13 | 2455 | |||
GenBank mRNA | BC085932.1 | - | 13 | 2352 | 7112012 |