Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nol10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008421.1 | NP_001008421.1 | 20 | 1989 | ||
GenBank mRNA | AK148210.1 | - | 20 | 2005 | ||
AK160864.1 | - | 20 | 2025 | |||
AK166160.1 | - | 20 | 2019 | |||
BC086676.1 | - | 20 | 1989 | 30659737 |