Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008502.2 | NP_001008502.2 | 2 | 1256 | ||
NM_001255992.1 | NP_001242921.1 | 2 | 1170 | |||
GenBank mRNA | AK156806.1 | - | 2 | 1171 | ||
BC085509.1 | - | 1 | 465 |