Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008558.1 | NP_001008558.1 | 1 | 399 | ||
GenBank mRNA | BC087630.1 | - | 1 | 399 | 7319134 |