Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008767.1 | NP_001008767.1 | 8 | 1820 | ||
GenBank mRNA | BC088411.1 | - | 8 | 1820 | 7192327 |