Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008771.3 | NP_001008771.2 | 10 | 1076 | ||
GenBank mRNA | AY321348.1 | - | 11 | 1037 | ||
BC097367.1 | - | 10 | 1027 | 7443204 | ||
FQ213175.1 | - | 10 | 1171 | |||
FQ226328.1 | - | 10 | 1071 |
Gene Symbol: LOC100911453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749373.3 | XP_003749421.1 | 10 | 1182 |