Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vom1r47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008916.1 | NP_001008916.1 | 1 | 378 | ||
GenBank mRNA | AY510316.1 | - | 1 | 378 | ||
AY510317.1 | - | 1 | 378 |