Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008973.2 | NP_001008973.2 | 10, 11 | 1233 | ||
NM_001289485.1 | NP_001276414.1 | 11, 12 | 1318 | |||
NM_001289486.1 | NP_001276415.1 | 10, 11 | 1230 | |||
GenBank mRNA | AF338468.1 | - | 8, 9 | 997 | ||
AF338469.1 | - | 11, 12 | 1312 | |||
AK053319.1 | - | 4, 5 | 567 | |||
AK135727.1 | - | 2, 3 | 224 | |||
BC050244.1 | - | 8, 9 | 1018 | |||
BC150885.1 | - | 10, 11 | 1195 | 9087796 | ||
BC172032.1 | - | 10, 11 | 1192 | 9054318 | ||
DV662151.1 | - | 2, 3 | 193 |