Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capza2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009180.2 | NP_001009180.1 | 2 | 117 | ||
GenBank mRNA | BC101867.1 | - | 2 | 117 | 7938377 |