Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009430.2 | NP_001009430.2 | 2 | 261 | ||
XM_006241250.2 | XP_006241312.1 | 2 | 430 | |||
GenBank mRNA | BC088210.1 | - | 2 | 334 | 7382451 |