Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3D19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009555.3 | NP_001009555.3 | 11 | 2257 | ||
NM_001128923.1 | NP_001122395.1 | 11 | 2245 | |||
NM_001128924.1 | NP_001122396.1 | 4 | 1248 | |||
NM_001243349.1 | NP_001230278.1 | 11 | 2166 | |||
XM_005262767.2 | XP_005262824.1 | 12 | 2118 | |||
XM_011531646.1 | XP_011529948.1 | 10 | 1904 | |||
XM_011531647.1 | XP_011529949.1 | 9 | 1796 | |||
XM_011531648.1 | XP_011529950.1 | 12 | 2231 | |||
XM_011531649.1 | XP_011529951.1 | 9 | 1796 | |||
XM_017007778.1 | XP_016863267.1 | 12 | 2222 | |||
XM_017007779.1 | XP_016863268.1 | 9 | 1787 | |||
XM_017007780.1 | XP_016863269.1 | 12 | 2222 | |||
XM_017007781.1 | XP_016863270.1 | 11 | 2148 | |||
XM_017007782.1 | XP_016863271.1 | 11 | 2148 | |||
XM_017007783.1 | XP_016863272.1 | 12 | 2118 | |||
GenBank mRNA | AK092528.1 | - | 4 | 478 | ||
AK128493.1 | - | 4 | 1097 | |||
AK291025.1 | - | 3 | 345 | |||
AK294476.1 | - | 11 | 2166 | |||
AL133047.1 | - | 1 | 21 | |||
BC010110.2 | - | 4 | 519 | |||
BC085613.1 | - | 2 | 236 | |||
BC108890.1 | - | 5 | 1356 | 40001764 | ||
BC108891.1 | - | 5 | 1356 | 40001765 | ||
BC108892.1 | - | 5 | 1356 | 40001766 | ||
BC108893.1 | - | 4 | 1248 | 40001767 | ||
BX647422.1 | - | 11 | 2169 |