Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIBP2 SIRNA Assays ›
Gene Symbol:
PPFIBP2
Gene Name:
PPFIA binding protein 2
Gene Aliases:
Cclp1
Chromosome Location:
Chr.11: 7513765 - 7667329 on Build GRCh38
UniGene ID:
Hs.655714
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219397
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256568.1
NP_001243497.1
16, 17
1717
NM_001256569.1
NP_001243498.1
15, 16
1697
NM_003621.3
NP_003612.2
19, 20
2294
XM_005253177.2
XP_005253234.1
19, 20
2289
XM_006718349.2
XP_006718412.1
20, 21
2322
XM_006718350.2
XP_006718413.1
20, 21
2322
XM_006718351.2
XP_006718414.1
18, 19
2130
XM_006718352.2
XP_006718415.1
17, 18
2097
XM_006718353.2
XP_006718416.1
16, 17
1689
XM_011520413.2
XP_011518715.1
21, 22
2310
XM_011520414.2
XP_011518716.1
20, 21
2037
XM_011520415.2
XP_011518717.1
20, 21
2322
XM_011520416.2
XP_011518718.1
20, 21
2322
XM_011520417.2
XP_011518719.1
18, 19
2177
XM_011520418.2
XP_011518720.1
17, 18
1774
XM_011520419.2
XP_011518721.1
18, 19
1830
XM_011520420.2
XP_011518722.1
18, 19
1773
XM_017018443.1
XP_016873932.1
20, 21
2277
XM_017018444.1
XP_016873933.1
18, 19
2130
XM_017018445.1
XP_016873934.1
17, 18
2144
XM_017018446.1
XP_016873935.1
17, 18
2097
XM_017018447.1
XP_016873936.1
16, 17
3390
GenBank mRNA
AF034803.1
-
16, 17
1622
AK001131.1
-
3, 4
223
AK025337.1
-
15, 16
1616
AK294554.1
-
8, 9
836
AK296718.1
-
16, 17
1717
AK301978.1
-
17, 18
1714
AK308398.1
-
19, 20
2060
AK311048.1
-
9, 10
863
AL834426.1
-
18, 19
1923
BC021026.1
-
2, 3
265
BC021714.1
-
19, 20
2242
4826610
CR749258.1
-
17, 18
1731
Back To Top
More Information
Gene Ontology Categories:
Process(es)
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-sshrn:80/10.239.165.44:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0