Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014728.3 | NP_055543.2 | 17 | 4785 | ||
XM_005274632.3 | XP_005274689.2 | 17 | 4458 | |||
XM_011545613.2 | XP_011543915.1 | 17 | 4767 | |||
XM_017029983.1 | XP_016885472.1 | 17 | 4670 | |||
XM_017029984.1 | XP_016885473.1 | 16 | 4369 | |||
XM_017029985.1 | XP_016885474.1 | 17 | 4461 | |||
XM_017029986.1 | XP_016885475.1 | 17 | 4674 | |||
GenBank mRNA | AB002314.3 | - | 17 | 4785 |