Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF518A SIRNA Assays ›
Gene Symbol:
ZNF518A
Gene Name:
zinc finger protein 518A
Gene Aliases:
ZNF518
Chromosome Location:
Chr.10: 96129220 - 96165512 on Build GRCh38
UniGene ID:
Hs.600823
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219445
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF518A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278524.1
NP_001265453.1
7
5771
NM_001278525.1
NP_001265454.1
6
5432
NM_001278526.1
NP_001265455.1
7
4056
NM_014803.3
NP_055618.2
6
5725
XM_011540406.2
XP_011538708.1
7
5584
XM_011540408.2
XP_011538710.1
6
6158
XM_011540410.2
XP_011538712.1
7
5606
XM_011540412.2
XP_011538714.1
6
6119
XM_011540413.2
XP_011538715.1
7
6244
XM_011540415.2
XP_011538717.1
6
5437
XM_011540418.2
XP_011538720.1
7
5553
XM_011540419.2
XP_011538721.1
5
5382
XM_011540420.2
XP_011538722.1
5
5409
XM_017016986.1
XP_016872475.1
7
6095
XM_017016987.1
XP_016872476.1
7
6122
XM_017016988.1
XP_016872477.1
7
6165
XM_017016989.1
XP_016872478.1
6
5460
XM_017016990.1
XP_016872479.1
6
5971
XM_017016991.1
XP_016872480.1
6
5944
XM_017016992.1
XP_016872481.1
7
5974
XM_017016993.1
XP_016872482.1
6
5455
XM_017016994.1
XP_016872483.1
6
6093
XM_017016995.1
XP_016872484.1
7
5998
XM_017016996.1
XP_016872485.1
7
6091
XM_017016997.1
XP_016872486.1
3
6304
XM_017016998.1
XP_016872487.1
2
6633
XM_017016999.1
XP_016872488.1
3
6086
GenBank mRNA
AB002333.2
-
2
5699
CR627437.1
-
6
5726
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0