Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194998.1 | NP_001181927.1 | 11 | 1567 | ||
NM_014985.3 | NP_055800.2 | 11 | 1567 | |||
XM_006720437.3 | XP_006720500.1 | 11 | 1752 | |||
XM_011521373.2 | XP_011519675.1 | 11 | 2320 | |||
XM_011521374.2 | XP_011519676.1 | 11 | 2319 | |||
XM_011521375.2 | XP_011519677.1 | 11 | 2319 | |||
XM_011521378.2 | XP_011519680.1 | 11 | 2319 | |||
XM_011521379.2 | XP_011519681.1 | 11 | 2318 | |||
XM_017022014.1 | XP_016877503.1 | 4 | 513 | |||
XM_017022016.1 | XP_016877505.1 | 11 | 2322 | |||
GenBank mRNA | AB020719.1 | - | 10 | 1172 | ||
AK303805.1 | - | 11 | 1490 | |||
BC069186.1 | - | 11 | 1463 | |||
BC107721.1 | - | 11 | 1571 | |||
BC117182.1 | - | 11 | 1408 | 40125733 | ||
BX648822.1 | - | 11 | 1494 |