Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020782.1 | NP_065833.1 | 3 | 1732 | ||
XM_017019698.1 | XP_016875187.1 | 3 | 2497 | |||
GenBank mRNA | AB037761.1 | - | 3 | 1463 | ||
AK095405.1 | - | 3 | 1732 | |||
BC028742.1 | - | 4 | 1600 | |||
BC137116.1 | - | 3 | 1698 | 9021113 | ||
BC137117.1 | - | 3 | 1698 | 9021114 |