Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020805.2 | NP_065856.1 | 2, 3 | 1340 | ||
XM_011526109.2 | XP_011524411.2 | 2, 3 | 1032 | |||
GenBank mRNA | AB037805.1 | - | 2, 3 | 1340 | ||
AK294996.1 | - | 1, 2 | 578 | |||
BC021267.2 | - | 2, 3 | 1208 | 4588702 |