Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020807.2 | NP_065858.1 | 2 | 3995 | ||
XM_005256069.3 | XP_005256126.1 | 2 | 3920 | |||
GenBank mRNA | AB037809.1 | - | 2 | 3995 |