Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286581.1 | NP_001273510.1 | 18 | 5039 | ||
NM_001286582.1 | NP_001273511.1 | 18 | 5033 | |||
NM_001286583.1 | NP_001273512.1 | 18 | 5027 | |||
NM_020901.3 | NP_065952.2 | 18 | 5036 | |||
XM_005253025.4 | XP_005253082.1 | 18 | 4997 | |||
XM_005253027.3 | XP_005253084.1 | 18 | 4985 | |||
XM_011520236.2 | XP_011518538.1 | 18 | 5033 | |||
XM_011520237.2 | XP_011518539.1 | 17 | 4880 | |||
GenBank mRNA | AB040975.1 | - | 18 | 4999 | ||
BC004950.1 | - | 5 | 891 | |||
BC013381.2 | - | 5 | 1515 | |||
BC029651.1 | - | 5 | 3165 | |||
BC041631.1 | - | 5 | 1426 | |||
BC112931.1 | - | 16 | 6769 | |||
BC127876.1 | - | 2 | 225 | |||
BC136615.1 | - | 18 | 4964 | 9020605 | ||
BC144293.1 | - | 18 | 4961 | 9052813 | ||
BC144294.1 | - | 18 | 4952 | 9052814 | ||
BC144295.1 | - | 18 | 4958 | 9052815 | ||
BC146771.1 | - | 18 | 4999 | 9007218 |