Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028821.3 | NP_083097.2 | 8 | 641 | ||
GenBank mRNA | AK005826.1 | - | 8 | 727 | ||
BC070454.1 | - | 598 | ||||
BC125392.1 | - | 7 | 623 | 40129907 | ||
BC125394.1 | - | 7 | 619 | 40129909 |