Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080527.1 | NP_001073996.1 | 33 | 4489 | ||
XM_006712539.3 | XP_006712602.1 | 34 | 4680 | |||
XM_011511218.2 | XP_011509520.1 | 34 | 4689 | |||
XM_017004169.1 | XP_016859658.1 | 33 | 4746 | |||
GenBank mRNA | AB290182.1 | - | 9 | 1343 | ||
AK074183.1 | - | 8 | 1421 | |||
BC035615.2 | - | 2 | 101 | |||
CR627350.1 | - | 2210 |