Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002815.2 | NP_001002815.2 | 9, 10 | 1152 | ||
XM_006248573.2 | XP_006248635.1 | 10, 11 | 1651 | |||
XM_008768824.1 | XP_008767046.1 | 9, 10 | 1395 | |||
XM_008768825.1 | XP_008767047.1 | 9, 10 | 1139 |