Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077440.1 | NP_001070908.1 | 12 | 4988 | ||
NM_001077441.1 | NP_001070909.1 | 13 | 4622 | |||
NM_001301038.1 | NP_001287967.1 | 13 | 5135 | |||
NM_014739.2 | NP_055554.1 | 13 | 5141 | |||
XM_005267237.3 | XP_005267294.1 | 12 | 4930 | |||
XM_017011529.1 | XP_016867018.1 | 13 | 4609 | |||
XM_017011530.1 | XP_016867019.1 | 12 | 5269 | |||
XM_017011531.1 | XP_016867020.1 | 12 | 4405 | |||
XM_017011532.1 | XP_016867021.1 | 14 | 5304 | |||
GenBank mRNA | BX537576.1 | - | 1 | 4340 | ||
BX648431.1 | - | 5 | 2804 | |||
BX648878.1 | - | 9 | 3882 | |||
D79986.1 | - | 13 | 5141 |