Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22003
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
7, 8
1150
NM_001205119.1
NP_001192048.1
6, 7
1094
NM_001205120.1
NP_001192049.1
6, 7
1094
NM_001205121.1
NP_001192050.1
7, 8
1145
NM_001205122.1
NP_001192051.1
5, 6
1012
NM_014741.4
NP_055556.2
7, 8
1150
XM_005253262.2
XP_005253319.1
7, 8
890
XM_005253263.2
XP_005253320.1
7, 8
1181
XM_005253265.2
XP_005253322.1
8, 9
1387
XM_005253266.2
XP_005253323.1
7, 8
1116
XM_005253268.2
XP_005253325.1
5, 6
493
XM_006718394.2
XP_006718457.1
8, 9
1237
XM_006718395.2
XP_006718458.1
8, 9
1172
XM_006718396.2
XP_006718459.1
7, 8
1271
XM_011520493.1
XP_011518795.1
9, 10
1374
XM_011520496.2
XP_011518798.1
6, 7
1149
XM_011520499.2
XP_011518801.1
6, 7
763
XM_011520500.1
XP_011518802.1
8, 9
1237
XM_011520501.1
XP_011518803.1
8, 9
1237
XM_011520502.1
XP_011518804.1
8, 9
1237
XM_011520503.1
XP_011518805.1
8, 9
1237
XM_017018599.1
XP_016874088.1
8, 9
1322
XM_017018600.1
XP_016874089.1
7, 8
890
XM_017018601.1
XP_016874090.1
6, 7
1065
XM_017018602.1
XP_016874091.1
8, 9
1387
XM_017018603.1
XP_016874092.1
9, 10
1504
XM_017018604.1
XP_016874093.1
7, 8
1181
XM_017018605.1
XP_016874094.1
8, 9
1172
XM_017018606.1
XP_016874095.1
7, 8
1116
XM_017018607.1
XP_016874096.1
5, 6
493
XM_017018608.1
XP_016874097.1
7, 8
890
XM_017018609.1
XP_016874098.1
7, 8
1181
XM_017018610.1
XP_016874099.1
6, 7
1065
XM_017018611.1
XP_016874100.1
7, 8
1116
XM_017018612.1
XP_016874101.1
8, 9
1387
XM_017018613.1
XP_016874102.1
7, 8
1181
XM_017018614.1
XP_016874103.1
8, 9
1172
XM_017018615.1
XP_016874104.1
7, 8
1271
XM_017018616.1
XP_016874105.1
6, 7
1065
XM_017018617.1
XP_016874106.1
8, 9
1270
XM_017018618.1
XP_016874107.1
8, 9
1237
XM_017018619.1
XP_016874108.1
7, 8
1121
XM_017018620.1
XP_016874109.1
7, 8
1181
XM_017018621.1
XP_016874110.1
6, 7
1065
XM_017018622.1
XP_016874111.1
7, 8
1133
GenBank mRNA
AB014552.1
-
6, 7
774
AK023867.1
-
7, 8
883
AK223603.1
-
6, 7
770
AK289641.1
-
7, 8
697
AK293109.1
-
7, 8
849
AK294110.1
-
5, 6
725
BC001331.1
-
7, 8
579
3458359
BC002378.2
-
7, 8
849
2961127
BC006191.2
-
7, 8
891
3936851
CD675643.1
-
5, 6
469
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0