Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002852.1 | NP_001002852.1 | 1, 2 | 251 | ||
XM_006253095.2 | XP_006253157.1 | 1, 2 | 256 | |||
GenBank mRNA | AY653229.1 | - | 1, 2 | 251 |