Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002897.3 | NP_001002897.2 | 14 | 3401 | ||
GenBank mRNA | AK010032.1 | - | 2 | 541 | ||
AY495541.1 | - | 14 | 3401 | |||
AY515312.1 | - | 9 | 1574 | |||
AY515313.1 | - | 14 | 3401 |