Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trappc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003708.1 | NP_001003708.1 | 5 | 596 | ||
GenBank mRNA | AY373378.1 | - | 5 | 596 | ||
BC098628.1 | - | 5 | 603 | 7311789 | ||
FQ220285.1 | - | 5 | 619 | |||
FQ229310.1 | - | 5 | 626 |