Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013910.2 | NP_038938.1 | 9 | 1896 | ||
GenBank mRNA | AF176524.2 | - | 2 | 317 | ||
AK031626.1 | - | 9 | 1862 | |||
AK034033.1 | - | 3 | 890 | |||
AK129479.1 | - | 20 | 3333 | |||
BC057622.1 | - | 9 | 1891 | 6402440 | ||
BC065090.1 | - | 20 | 3532 | 6851421 |