Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gspt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003978.1 | NP_001003978.1 | 8 | 1241 | ||
GenBank mRNA | BC079092.1 | - | 8 | 1241 | 7123818 |
Gene Symbol: LOC100911685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750763.3 | XP_003750811.1 | 1664 | |||
XM_006245740.2 | XP_006245802.1 | 1667 |