Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136264.1 | NP_001129736.1 | 5 | 847 | ||
NM_001136266.1 | NP_001129738.1 | 4 | 836 | |||
NM_001136267.1 | NP_001129739.1 | 4 | 817 | |||
NM_001136268.1 | NP_001129740.1 | 3 | 667 | |||
NM_014764.3 | NP_055579.1 | 4 | 913 | |||
GenBank mRNA | AK095963.1 | - | 5 | 706 | ||
AK125855.1 | - | 2 | 2560 | |||
AK290119.1 | - | 4 | 813 | |||
AK303828.1 | - | 4 | 736 | |||
AY430097.1 | - | 4 | 812 | |||
BC002334.2 | - | 4 | 795 | 3509229 | ||
BC007900.2 | - | 4 | 794 | 4297784 | ||
BX647278.1 | - | 4 | 811 | |||
D31767.1 | - | 4 | 798 |