Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004102.1 | NP_001004102.1 | 8 | 990 | ||
XM_006236132.2 | XP_006236194.1 | 9 | 1899 | |||
XM_006236133.2 | XP_006236195.1 | 9 | 2192 | |||
XM_006236134.2 | XP_006236196.1 | 9 | 1428 | |||
XM_006236135.2 | XP_006236197.1 | 8 | 1359 | |||
XM_008762700.1 | XP_008760922.1 | 9 | 2624 | |||
XM_008762701.1 | XP_008760923.1 | 10 | 1586 | |||
GenBank mRNA | BC078945.1 | - | 10 | 1246 | ||
BC099820.1 | - | 8 | 1072 | 7930680 |