Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004143.4 | NP_001004143.2 | 13 | 2374 | ||
GenBank mRNA | AK034492.1 | - | 13 | 2349 | ||
AK084022.1 | - | 9 | 1533 | |||
AK155071.1 | - | 13 | 2375 | |||
AK160992.1 | - | 13 | 2283 | |||
AK220368.1 | - | 13 | 2335 | |||
BC026701.1 | - | 6 | 1146 | |||
BC052540.1 | - | 9 | 1668 | |||
BC058419.1 | - | 6 | 1193 | |||
BC080737.1 | - | 13 | 2245 | 30431735 |