Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270934.1 | NP_001257863.1 | 21 | 3123 | ||
NM_001270941.1 | NP_001257870.1 | 22 | 3186 | |||
NM_001282282.1 | NP_001269211.1 | 21 | 2909 | |||
NM_014790.4 | NP_055605.2 | 21 | 3115 | |||
XM_017010097.1 | XP_016865586.1 | 23 | 3180 | |||
XM_017010098.1 | XP_016865587.1 | 21 | 3066 | |||
GenBank mRNA | AB011127.1 | - | 21 | 2817 | ||
AF273057.1 | - | 21 | 2956 | |||
AK291280.1 | - | 21 | 2837 | |||
AK299724.1 | - | 21 | 2835 | |||
BC017354.2 | - | 21 | 2880 | 5020741 |