Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098815.2 | NP_001092285.1 | 11 | 2751 | ||
NM_001136030.2 | NP_001129502.1 | 11 | 2848 | |||
NM_001261844.1 | NP_001248773.1 | 10 | 2600 | |||
NM_014796.2 | NP_055611.1 | 9 | 2691 | |||
XM_005269247.2 | XP_005269304.1 | 10 | 2674 | |||
XM_006719715.3 | XP_006719778.1 | 13 | 3182 | |||
XM_011539035.2 | XP_011537337.1 | 13 | 3195 | |||
XM_011539036.1 | XP_011537338.1 | 12 | 3016 | |||
XM_011539037.2 | XP_011537339.1 | 11 | 7236 | |||
XM_017020262.1 | XP_016875751.1 | 13 | 4245 | |||
XM_017020263.1 | XP_016875752.1 | 13 | 4151 | |||
GenBank mRNA | AB018291.1 | - | 9 | 2885 |