Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004210.2 | NP_001004210.1 | 5 | 919 | ||
NM_001271731.1 | NP_001258660.1 | 6 | 893 | |||
GenBank mRNA | BC079450.1 | - | 5 | 900 | 7115933 | |
FQ225483.1 | - | 1 | 126 | |||
FQ232289.1 | - | 1 | 126 |