Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004240.1 | NP_001004240.1 | 3 | 392 | ||
GenBank mRNA | BC078984.1 | - | 3 | 392 | 7112302 |