Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004263.1 | NP_001004263.1 | 14 | 2139 | ||
XM_006234232.1 | XP_006234294.1 | 16 | 2482 | |||
XM_006234233.2 | XP_006234295.1 | 14 | 2388 | |||
XM_008761845.1 | XP_008760067.1 | 16 | 2309 | |||
XM_008761846.1 | XP_008760068.1 | 14 | 2286 | |||
GenBank mRNA | BC079069.1 | - | 14 | 2139 | 7121642 |