Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004266.1 | NP_001004266.1 | 7 | 559 | ||
XM_006233794.2 | XP_006233856.1 | 8 | 595 | |||
GenBank mRNA | BC079446.1 | - | 7 | 559 | 7115585 |