Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004279.1 | NP_001004279.1 | 3 | 348 | ||
GenBank mRNA | BC076386.1 | - | 3 | 348 | 7108860 | |
FQ214826.1 | - | 3 | 351 |
Gene Symbol: Ppidl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750757.3 | XP_003750805.1 | 1 | 296 |